siRNA Details
Gene Symbol: Mark1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053947.1 | NP_446399.1 | 2 | 241 | ||
XM_008769816.1 | XP_008768038.1 | 3 | 245 | |||
GenBank mRNA | Z83868.1 | - | 2 | 241 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
