siRNA Details
Gene Symbol: Mark1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053947.1 | NP_446399.1 | 16 | 2111 | ||
XM_008769816.1 | XP_008768038.1 | 17 | 2118 | |||
GenBank mRNA | Z83868.1 | - | 16 | 2111 |

More Information
Other Species Targeted

Gene Ontology Categories:
