siRNA Details
Gene Symbol: Ccr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053958.1 | NP_446410.1 | 3 | 1107 | ||
XM_006244178.2 | XP_006244240.1 | 3 | 1885 | |||
GenBank mRNA | Y13400.1 | - | 3 | 1107 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
