siRNA Details
Gene Symbol: Eif4e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053974.2 | NP_446426.1 | 7 | 564 | ||
XM_008761488.1 | XP_008759710.1 | 8 | 677 | |||
GenBank mRNA | BC087001.1 | - | 7 | 564 | 7122606 | |
FQ218941.1 | - | 7 | 556 | |||
FQ225918.1 | - | 2 | 56 | |||
X83399.1 | - | 7 | 599 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_007917, AK146757, BC010759, M61731, AK167097, and others.
Gene Ontology Categories:
