siRNA Details
Gene Symbol: Kcnj12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053981.2 | NP_446433.2 | 2 | 487 | ||
XM_008767763.1 | XP_008765985.1 | 2 | 7777 | |||
GenBank mRNA | AY376691.1 | - | 1 | 155 | ||
BC127487.1 | - | 2 | 487 | 8364361 | ||
X78461.1 | - | 2 | 218 |

More Information
Other Species Targeted

Gene Ontology Categories:
