siRNA Details
Gene Symbol: Rab8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053998.2 | NP_446450.2 | 6, 7 | 489 | ||
GenBank mRNA | BC071176.1 | - | 6, 7 | 482 | ||
BC087584.1 | - | 5, 6 | 320 | |||
BC105863.1 | - | 6, 7 | 489 | 7457017 | ||
JN019799.1 | - | 6, 7 | 484 | |||
M83675.1 | - | 4, 5 | 268 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
