siRNA Details
Gene Symbol: Ppp2r2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053999.2 | NP_446451.1 | 10 | 1452 | ||
XM_008770730.1 | XP_008768952.1 | 10 | 2259 | |||
XM_008770731.1 | XP_008768953.1 | 10 | 2259 | |||
XM_008770732.1 | XP_008768954.1 | 8 | 1041 | |||
GenBank mRNA | BC128731.1 | - | 10 | 1452 | 7370343 | |
M83297.1 | - | 11 | 1398 | |||
M83298.1 | - | 10 | 1433 |
