siRNA Details
Gene Symbol: Ctrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054009.1 | NP_446461.1 | 2 | 85 | ||
GenBank mRNA | AB020757.1 | - | 2 | 85 | ||
FQ234836.1 | - | 2 | 97 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
