siRNA Details
Gene Symbol: Sh3bp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054011.1 | NP_446463.2 | 3 | 252 | ||
GenBank mRNA | AB027562.1 | - | 3 | 252 | ||
FQ210473.1 | - | 3 | 379 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
