siRNA Details
Gene Symbol: Slc15a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079838.1 | NP_001073307.1 | 2 | 315 | ||
NM_057121.1 | NP_476462.1 | 22 | 1950 | |||
XM_006252470.2 | XP_006252532.1 | 21 | 2191 | |||
GenBank mRNA | AY860424.1 | - | 2 | 315 | ||
D50306.1 | - | 22 | 1948 | |||
D50664.1 | - | 22 | 1950 |
