siRNA Details
Gene Symbol: Slc26a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057127.1 | NP_476468.1 | 5 | 2245 | ||
XM_006254784.2 | XP_006254846.1 | 3 | 4221 | |||
XM_006254785.2 | XP_006254847.1 | 4 | 2891 | |||
XM_006254786.2 | XP_006254848.1 | 4 | 3699 | |||
XM_006254787.2 | XP_006254849.1 | 4 | 2239 | |||
XM_006254788.2 | XP_006254850.1 | 3 | 2104 | |||
GenBank mRNA | D82883.1 | - | 5 | 2245 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
