siRNA Details
Gene Symbol: Hnrnpk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057141.1 | NP_476482.1 | 14 | 1254 | ||
XM_006253552.2 | XP_006253614.1 | 16 | 1661 | |||
XM_006253553.2 | XP_006253615.1 | 15 | 1357 | |||
XM_006253554.2 | XP_006253616.1 | 16 | 1661 | |||
XM_006253555.2 | XP_006253617.1 | 15 | 1589 | |||
GenBank mRNA | BC061867.1 | - | 15 | 1341 | 5598872 | |
D17711.1 | - | 14 | 1254 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_025279, BC006694, AK128891, AK011428, AK152927, and others.
Gene Ontology Categories:
