siRNA Details
Gene Symbol: Oxr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197907.1 | NP_001184836.1 | 5 | 583 | ||
NM_057153.3 | NP_476494.2 | 3 | 265 | |||
GenBank mRNA | BC166763.1 | - | 3 | 481 | 7931847 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
