siRNA Details
Gene Symbol: Ccl22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057203.1 | NP_476551.1 | 2 | 170 | ||
GenBank mRNA | AF163477.1 | - | 2 | 115 | ||
AF432871.2 | - | 2 | 147 | |||
BC088218.1 | - | 2 | 170 | 7384271 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
