siRNA Details
Gene Symbol: Ptpn23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057204.2 | NP_476552.2 | 18, 19 | 1987 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001081043, BC059902.
Gene Ontology Categories:
