siRNA Details
Gene Symbol: Sf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110793.1 | NP_001104263.1 | 4 | 684 | ||
NM_058210.2 | NP_478117.2 | 4 | 684 | |||
XM_006230641.2 | XP_006230703.1 | 4 | 3585 | |||
XM_006230642.2 | XP_006230704.1 | 4 | 3585 | |||
XM_006230643.2 | XP_006230705.1 | 4 | 3585 | |||
XM_006230644.2 | XP_006230706.1 | 4 | 3584 | |||
XM_006230645.2 | XP_006230707.1 | 4 | 3584 | |||
XM_006230646.2 | XP_006230708.1 | 3 | 3509 | |||
XM_006230647.2 | XP_006230709.1 | 4 | 3583 | |||
XM_006230648.2 | XP_006230710.1 | 4 | 3583 | |||
XM_008760065.1 | XP_008758287.1 | 2 | 1498 | |||
XM_008760066.1 | XP_008758288.1 | 4 | 3585 | |||
GenBank mRNA | BC081859.1 | - | 3 | 597 | 7132476 | |
FQ224273.1 | - | 4 | 687 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
