siRNA Details
Gene Symbol: Rps20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007603.2 | NP_001007604.1 | 4 | 398 | ||
GenBank mRNA | BC058496.1 | - | 4 | 402 | 6891327 | |
FQ210377.1 | - | 4 | 399 | |||
FQ210861.1 | - | 4 | 398 | |||
FQ210956.1 | - | 4 | 398 | |||
FQ221135.1 | - | 4 | 398 | |||
FQ221327.1 | - | 4 | 401 | |||
FQ221535.1 | - | 4 | 398 | |||
FQ221861.1 | - | 4 | 402 | |||
FQ222030.1 | - | 4 | 400 | |||
FQ222128.1 | - | 4 | 399 | |||
FQ222290.1 | - | 4 | 398 | |||
FQ222476.1 | - | 4 | 398 | |||
FQ222864.1 | - | 4 | 398 | |||
FQ222926.1 | - | 4 | 399 | |||
FQ224107.1 | - | 4 | 398 | |||
FQ224441.1 | - | 4 | 398 | |||
FQ228423.1 | - | 4 | 402 | |||
FQ228564.1 | - | 4 | 398 | |||
FQ229018.1 | - | 4 | 401 | |||
X51537.1 | - | 4 | 380 |
Gene Symbol: LOC100359951 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729994.4 | XP_002730040.1 | 1 | 400 |
Gene Symbol: LOC100362149 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729158.4 | XP_002729204.1 | 1 | 408 |
Gene Symbol: LOC100362684 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729574.4 | XP_002729620.1 | 1 | 398 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
