siRNA Details
Gene Symbol: Ackr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_078621.2 | NP_511176.2 | 1 | 686 | ||
XM_008766664.1 | XP_008764886.1 | 3 | 1204 | |||
XM_008766665.1 | XP_008764887.1 | 3 | 1545 | |||
GenBank mRNA | BC086449.1 | - | 1 | 686 | 7304127 | |
U92803.1 | - | 2 | 552 |
