siRNA Details
Gene Symbol: Pcyt1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_078622.2 | NP_511177.2 | 8 | 841 | ||
XM_006248444.2 | XP_006248506.1 | 8 | 994 | |||
XM_008768694.1 | XP_008766916.1 | 8 | 1192 | |||
GenBank mRNA | BC085713.1 | - | 8 | 839 | 7127059 | |
L13245.1 | - | 7 | 738 | |||
M36071.1 | - | 8 | 838 | |||
U03490.1 | - | 8 | 803 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
