siRNA Details
Gene Symbol: Atp5i | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080481.1 | NP_536729.1 | 3 | 170 | ||
GenBank mRNA | BC058449.1 | - | 168 | 6890563 | ||
D13121.1 | - | 3 | 170 | |||
FQ223740.1 | - | 3 | 160 | |||
FQ224619.1 | - | 309 |

More Information
Other Species Targeted

Gene Ontology Categories:
