siRNA Details
Gene Symbol: Nploc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080577.2 | NP_542144.1 | 4 | 375 | ||
GenBank mRNA | AF234600.1 | - | 4 | 303 | ||
BC101887.1 | - | 4 | 375 | 7930738 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001195023, NM_199469, BC065156, AK149823, AK141998, and others.
Gene Ontology Categories:
