siRNA Details
Gene Symbol: Rplp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030021.1 | NP_001025192.1 | 2, 3 | 164 | ||
GenBank mRNA | BC099697.1 | - | 2, 3 | 164 | 7934014 | |
FQ217138.1 | - | 2, 3 | 177 | |||
FQ217658.1 | - | 2, 3 | 178 | |||
FQ221181.1 | - | 2, 3 | 177 | |||
FQ221478.1 | - | 2, 3 | 180 | |||
FQ221584.1 | - | 2, 3 | 176 | |||
FQ221984.1 | - | 2, 3 | 178 | |||
FQ222461.1 | - | 2, 3 | 176 | |||
FQ222498.1 | - | 2, 3 | 177 | |||
FQ223422.1 | - | 2, 3 | 179 | |||
FQ228678.1 | - | 2, 3 | 178 | |||
X15098.1 | - | 2, 3 | 178 |
Gene Symbol: LOC100911575 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749013.2 | XP_003749061.1 | 2, 3 | 399 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
