siRNA Details
Gene Symbol: Abcb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080582.1 | NP_542149.1 | 10, 11 | 1817 | ||
GenBank mRNA | AF106563.1 | - | 2, 3 | 204 | ||
AJ003004.1 | - | 10, 11 | 1817 | |||
BC085712.1 | - | 10, 11 | 1833 | 7190755 | ||
FQ211570.1 | - | 10, 11 | 1843 |
