siRNA Details
Gene Symbol: Ap2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080583.1 | NP_542150.1 | 11 | 1485 | ||
XM_008767935.1 | XP_008766157.1 | 12 | 2242 | |||
XM_008767936.1 | XP_008766158.1 | 11 | 1456 | |||
XM_008767937.1 | XP_008766159.1 | 10 | 1371 | |||
XM_008767938.1 | XP_008766160.1 | 10 | 1388 | |||
XM_008767939.1 | XP_008766161.1 | 11 | 2224 | |||
GenBank mRNA | BC103481.1 | - | 12 | 1454 | 7929959 | |
M34176.1 | - | 11 | 1417 | |||
M77246.1 | - | 11 | 1485 |
