siRNA Details
Gene Symbol: Phkg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080584.4 | NP_542151.1 | 2 | 489 | ||
XM_008759855.1 | XP_008758077.1 | 3 | 312 | |||
XM_008759856.1 | XP_008758078.1 | 3 | 430 | |||
GenBank mRNA | BC107906.1 | - | 2 | 413 | ||
BC128715.1 | - | 2 | 418 | 7456955 | ||
M73808.1 | - | 2 | 489 |
