siRNA Details
Gene Symbol: Plcd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080688.1 | NP_542419.1 | 13, 14 | 2076 | ||
XM_006245135.2 | XP_006245197.1 | 15, 16 | 3247 | |||
XM_006245136.2 | XP_006245198.1 | 14, 15 | 2088 | |||
XM_006245137.2 | XP_006245199.1 | 14, 15 | 2387 | |||
XM_006245138.2 | XP_006245200.1 | 14, 15 | 2262 | |||
XM_008767182.1 | XP_008765404.1 | 14, 15 | 2666 | |||
XM_008767183.1 | XP_008765405.1 | 14, 15 | 2666 | |||
XM_008767184.1 | XP_008765406.1 | 14, 15 | 2705 | |||
XM_008767185.1 | XP_008765407.1 | 14, 15 | 2089 | |||
XM_008767186.1 | XP_008765408.1 | 14, 15 | 2667 | |||
XM_008767187.1 | XP_008765409.1 | 14, 15 | 2664 | |||
XM_008767188.1 | XP_008765410.1 | 14, 15 | 2612 | |||
XM_008767189.1 | XP_008765411.1 | 14, 15 | 2666 | |||
XM_008767190.1 | XP_008765412.1 | 13, 14 | 2569 | |||
XM_008767191.1 | XP_008765413.1 | 13, 14 | 2566 | |||
GenBank mRNA | D50455.1 | - | 13, 14 | 2066 | ||
U16655.1 | - | 13, 14 | 2076 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
