siRNA Details
Gene Symbol: Plcd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080688.1 | NP_542419.1 | 13 | 2057 | ||
XM_006245135.2 | XP_006245197.1 | 15 | 3228 | |||
XM_006245136.2 | XP_006245198.1 | 14 | 2069 | |||
XM_006245137.2 | XP_006245199.1 | 14 | 2368 | |||
XM_006245138.2 | XP_006245200.1 | 14 | 2243 | |||
XM_008767182.1 | XP_008765404.1 | 14 | 2647 | |||
XM_008767183.1 | XP_008765405.1 | 14 | 2647 | |||
XM_008767184.1 | XP_008765406.1 | 14 | 2686 | |||
XM_008767185.1 | XP_008765407.1 | 14 | 2070 | |||
XM_008767186.1 | XP_008765408.1 | 14 | 2648 | |||
XM_008767187.1 | XP_008765409.1 | 14 | 2645 | |||
XM_008767188.1 | XP_008765410.1 | 14 | 2593 | |||
XM_008767189.1 | XP_008765411.1 | 14 | 2647 | |||
XM_008767190.1 | XP_008765412.1 | 13 | 2550 | |||
XM_008767191.1 | XP_008765413.1 | 13 | 2547 | |||
GenBank mRNA | D50455.1 | - | 13 | 2047 | ||
U16655.1 | - | 13 | 2057 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
