siRNA Details
Gene Symbol: Cacng6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080694.1 | NP_542425.1 | 2, 3 | 403 | ||
GenBank mRNA | AF361343.1 | - | 2, 3 | 403 |

More Information
Other Species Targeted

Gene Ontology Categories:
