siRNA Details
Gene Symbol: Cacng6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080694.1 | NP_542425.1 | 2 | 395 | ||
XM_008758749.1 | XP_008756971.1 | 2 | 1475 | |||
GenBank mRNA | AF361343.1 | - | 2 | 395 | ||
AF361344.1 | - | 2 | 395 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
