siRNA Details
Gene Symbol: Dynll2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080697.2 | NP_542428.1 | 3 | 473 | ||
XM_006247077.2 | XP_006247139.1 | 3 | 252 | |||
XM_008767940.1 | XP_008766162.1 | 3 | 243 | |||
GenBank mRNA | AY034383.1 | - | 3 | 367 | ||
BC061874.1 | - | 3 | 473 | 5598327 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001168471, NM_026556, NM_001168472, BC011289, AK028319, and others.
Gene Ontology Categories:
