siRNA Details
Gene Symbol: Dgka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080787.1 | NP_542965.1 | 23, 24 | 2636 | ||
XM_008765013.1 | XP_008763235.1 | 22, 23 | 3638 | |||
XM_008765019.1 | XP_008763241.1 | 23, 24 | 3415 | |||
XM_008765020.1 | XP_008763242.1 | 23, 24 | 3342 | |||
GenBank mRNA | FQ209812.1 | - | 23, 24 | 2530 | ||
S49760.1 | - | 23, 24 | 2636 |

More Information
Other Species Targeted

Gene Ontology Categories:
