siRNA Details
Gene Symbol: Dgka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080787.1 | NP_542965.1 | 16 | 1832 | ||
XM_008765012.1 | XP_008763234.1 | 15 | 2834 | |||
XM_008765013.1 | XP_008763235.1 | 15 | 2834 | |||
XM_008765014.1 | XP_008763236.1 | 15 | 2744 | |||
XM_008765015.1 | XP_008763237.1 | 15 | 2676 | |||
XM_008765016.1 | XP_008763238.1 | 15 | 1917 | |||
XM_008765017.1 | XP_008763239.1 | 16 | 2702 | |||
XM_008765018.1 | XP_008763240.1 | 16 | 2612 | |||
XM_008765019.1 | XP_008763241.1 | 16 | 2611 | |||
XM_008765020.1 | XP_008763242.1 | 16 | 2538 | |||
GenBank mRNA | FQ209812.1 | - | 16 | 1726 | ||
S49760.1 | - | 16 | 1832 |

More Information
Other Species Targeted

Gene Ontology Categories:
