siRNA Details
Gene Symbol: Il2rg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080889.1 | NP_543165.1 | 4, 5 | 612 | ||
GenBank mRNA | AF410926.1 | - | 4, 5 | 532 | ||
AY325195.1 | - | 440 | ||||
BC079343.1 | - | 4, 5 | 612 | 7106866 | ||
FQ232752.1 | - | 3, 4 | 413 |
