siRNA Details
Gene Symbol: Daxx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080891.2 | NP_543167.2 | 3 | 1088 | ||
XM_006256103.2 | XP_006256165.1 | 3 | 1870 | |||
XM_006256104.2 | XP_006256166.1 | 3 | 1544 | |||
XM_006256105.2 | XP_006256167.1 | 3 | 1440 | |||
XM_006256106.2 | XP_006256168.1 | 3 | 1106 | |||
XM_006256107.2 | XP_006256169.1 | 3 | 1210 | |||
XM_006256108.2 | XP_006256170.1 | 3 | 1090 | |||
XM_006256109.1 | XP_006256171.1 | 2 | 1963 | |||
XM_006256110.2 | XP_006256172.1 | 3 | 1089 | |||
XM_008772704.1 | XP_008770926.1 | 2 | 1963 | |||
GenBank mRNA | AB064671.1 | - | 2 | 992 | ||
BC128729.1 | - | 3 | 1088 | 8366304 | ||
FQ223571.1 | - | 3 | 1059 |
