siRNA Details
Gene Symbol: Pde11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127480.1 | NP_001120952.1 | 6 | 769 | ||
NM_001127481.2 | NP_001120953.1 | 4 | 373 | |||
NM_080893.1 | NP_543169.1 | 4 | 1447 | |||
GenBank mRNA | AB059360.1 | - | 4 | 375 | ||
AB059361.1 | - | 6 | 769 | |||
AB059362.1 | - | 4 | 1447 |
