siRNA Details
Gene Symbol: Pde11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080893.1 | NP_543169.1 | 1 | 243 | ||
GenBank mRNA | AB059362.1 | - | 1 | 243 |

More Information
Other Species Targeted

Gene Ontology Categories:
