siRNA Details
Gene Symbol: Pde11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127480.1 | NP_001120952.1 | 8 | 904 | ||
NM_001127481.2 | NP_001120953.1 | 6 | 508 | |||
NM_080893.1 | NP_543169.1 | 6 | 1582 | |||
GenBank mRNA | AB059360.1 | - | 6 | 510 | ||
AB059361.1 | - | 8 | 904 | |||
AB059362.1 | - | 6 | 1582 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
