siRNA Details
Gene Symbol: Bnip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080897.1 | NP_543173.1 | 6 | 567 | ||
GenBank mRNA | AF454391.1 | - | 6 | 567 | ||
FQ218385.1 | - | 6 | 571 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
