siRNA Details
Gene Symbol: Higd1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080902.4 | NP_543178.2 | 3 | 244 | ||
GenBank mRNA | AY062253.3 | - | 3 | 303 | ||
BC059118.1 | - | 3 | 248 | 6890532 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
