siRNA Details
Gene Symbol: Arf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080904.2 | NP_543180.1 | 1, 2 | 230 | ||
XM_006257320.2 | XP_006257382.1 | 2, 3 | 439 | |||
GenBank mRNA | AY325223.1 | - | 2, 3 | 197 | ||
BC088865.1 | - | 1, 2 | 230 | 7319260 | ||
L12382.1 | - | 2, 3 | 343 |

More Information
Other Species Targeted

Gene Ontology Categories:
