siRNA Details
Gene Symbol: Siah1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080905.2 | NP_543181.2 | 2 | 852 | ||
XM_006255211.2 | XP_006255273.1 | 2 | 5453 | |||
XM_006255213.2 | XP_006255275.1 | 2 | 1192 | |||
XM_008772345.1 | XP_008770567.1 | 3 | 893 | |||
GenBank mRNA | AF389476.1 | - | 1 | 672 | ||
AY681965.1 | - | 1 | 672 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
