siRNA Details
Gene Symbol: Rtn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009953.2 | NP_001009953.2 | 4 | 655 | ||
NM_080909.2 | NP_543185.2 | 5 | 2829 | |||
XM_006230649.2 | XP_006230711.1 | 6 | 2901 | |||
XM_006230650.2 | XP_006230712.1 | 4 | 625 | |||
GenBank mRNA | AF442357.1 | - | 4 | 659 | ||
AF442358.1 | - | 4 | 659 | |||
AY164698.1 | - | 4 | 659 | |||
AY496443.1 | - | 5 | 2829 | |||
BC062068.1 | - | 4 | 653 | |||
BC107448.1 | - | 4 | 655 | |||
BC129077.1 | - | 4 | 663 |
