siRNA Details
Gene Symbol: Ugt1a7c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130407.2 | NP_569091.2 | 260 | |||
GenBank mRNA | AF461738.1 | - | 268 | |||
AY435134.1 | - | 227 | ||||
BC078732.1 | - | 263 | ||||
U75903.1 | - | 269 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
