siRNA Details
Gene Symbol: Cfh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130409.2 | NP_569093.2 | 11 | 1606 | ||
XM_008769531.1 | XP_008767753.1 | 10 | 1632 | |||
XM_008769533.1 | XP_008767755.1 | 11 | 1803 | |||
GenBank mRNA | AJ320522.1 | - | 1580 |
