siRNA Details
Gene Symbol: Il23a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130410.2 | NP_569094.1 | 3 | 684 | ||
GenBank mRNA | AY055379.1 | - | 3 | 412 | ||
BC098907.1 | - | 3 | 684 | 7454618 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Il23a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130410.2 | NP_569094.1 | 3 | 684 | ||
GenBank mRNA | AY055379.1 | - | 3 | 412 | ||
BC098907.1 | - | 3 | 684 | 7454618 |