siRNA Details
Gene Symbol: Sdf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130412.2 | NP_569096.2 | 6 | 955 | ||
XM_006239516.2 | XP_006239578.1 | 6 | 1003 | |||
GenBank mRNA | AF405545.1 | - | 5 | 865 | ||
BC086996.1 | - | 6 | 955 | 7192272 | ||
FQ218523.1 | - | 6 | 981 | |||
FQ219323.1 | - | 6 | 983 | |||
FQ232476.1 | - | 6 | 981 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
