siRNA Details
Gene Symbol: Runx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130425.1 | NP_569109.1 | 3 | 463 | ||
GenBank mRNA | AF421886.1 | - | 3 | 463 | ||
FQ230848.1 | - | 212 |

More Information
Other Species Targeted

Gene Ontology Categories:
