siRNA Details
Gene Symbol: Cygb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130744.2 | NP_570100.1 | 2 | 525 | ||
GenBank mRNA | AJ245663.2 | - | 2 | 433 | ||
BC088455.1 | - | 2 | 525 | 7127197 |

More Information
Other Species Targeted

Gene Ontology Categories:
