siRNA Details
Gene Symbol: Slc16a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138831.1 | NP_620186.1 | 3 | 909 | ||
XM_008772974.1 | XP_008771196.1 | 3 | 948 | |||
XM_008772975.1 | XP_008771197.1 | 3 | 947 | |||
GenBank mRNA | AB047324.1 | - | 3 | 909 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
