siRNA Details
Gene Symbol: Slc16a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138831.1 | NP_620186.1 | 3, 4 | 1137 | ||
XM_008772974.1 | XP_008771196.1 | 3, 4 | 1176 | |||
GenBank mRNA | AB047324.1 | - | 3, 4 | 1137 |

More Information
Other Species Targeted

Gene Ontology Categories:
